Learn More
Hach Company Operator RFID Tag, PK/2
Supplier:  Hach Company LZQ066
Encompass
The operator tag allows the automatic operator recognition by the DR 3900 or DR 6000 photometer or LOC 100 sample identification device.
Catalog No. 50-199-7893