Learn More
Enzo Life Sciences Osteoprotegerin monoclonal antibody (98A1071) (100µg)
SDP
Supplier:  Enzo Life Sciences ADIAAM020E
Encompass_Preferred
Clone: 98A1071. Host: Mouse. Isotype: IgG1. Immunogen: Synthetic peptide corresponding to the sequence near the N-terminus of human OPG (osteoprotegerin). UniProt ID: O00300. Species reactivity: Human. Applications: WB. Recommended Dilutions: Western Blot (1µg/ml AP, 1µg/ml ECL) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein G affinity purified. Formulation: Liquid. In PBS containing 0.05% BSA and 0.05% sodium azide.
Catalog No. 50-200-7132