Learn More
Medchemexpress LLC HY-N6061 1mg Medchemexpress, Ophiopogonin C CAS:65586-25-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN60611MG
Encompass_Preferred
Medchemexpress, HY-N6061 1mg Ophiopogonin C CAS:65586-25-6 Ophiopogonin C, isolated from the tubers of Ophiopogon japonicus, is a rare naturally occurring C29 steroidal glycoside. Ophiopogonin C shows cytotoxic activity against two human tumor cell lines MG-63 and SNU387 with IC50s of 19.76 μM and 15.51 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0447