Learn More
Cayman Chemical Ophiopogonin D, Purity: ≥95%, CAS: 945619-74-9, 10 mg
SDP
Catalog No. 502948233
Encompass_Preferred
A steroid glycoside; induces apoptosis in, and reduces TGF-β1-induced migration of, MDA-MB-231 human breast cancer cells at 2.5, 5, or 10 µM; inhibits hydrogen peroxide-induced ROS production in RAW 264.7 cells at 10 and 100 µM; peritoneal neutrophil infiltration in a mouse model of zymosan A-induced peritonitis at 1 mg/kg; reduces thrombus weight in a mouse model of venous thrombosis at 1 and 2 mg/kg
Catalog No. 50-294-8233 Supplier Cayman Chemical Supplier No. 3438510 MG
A steroid glycoside; induces apoptosis in, and reduces TGF-β1-induced migration of, MDA-MB-231 human breast cancer cells at 2.5, 5, or 10 µM; inhibits hydrogen peroxide-induced ROS production in RAW 264.7 cells at 10 and 100 µM; peritoneal neutrophil infiltration in a mouse model of zymosan A-induced peritonitis at 1 mg/kg; reduces thrombus weight in a mouse model of venous thrombosis at 1 and 2 mg/kg