Learn More
Cayman Chemical Ophopogons radx Ruscogen 10mg
SDP
Catalog No. 502934427
Encompass_Preferred
A steroid sapogenin with diverse biological activities; reduces PMA-induced adhesion of HL-60 cells to ECV304 cells (IC50 = 7.76 nM); decreases zymosan A-induced peritoneal leukocyte migration in rats at 5 mg/kg; inhibits ADP-induced platelet aggregation in rats at 12.5 mg/kg; prevents monocrotaline-induced increases in MAP, expression of IL-1β and TNF in plasma and lung, and endothelial cell apoptosis in pulmonary arterioles in a rat model of pulmonary hypertension; reduces proteinuria and renal macrophage infiltration in a rat model of streptozotocin-induced diabetes; decreases gene expression of inflammatory cytokines, dyslipidemia, and liver steatosis in a hamster model of high-fat diet-induced NASH
Catalog No. 50-293-4427 Supplier Cayman Chemical Supplier No. 2521710 MG
A steroid sapogenin with diverse biological activities; reduces PMA-induced adhesion of HL-60 cells to ECV304 cells (IC50 = 7.76 nM); decreases zymosan A-induced peritoneal leukocyte migration in rats at 5 mg/kg; inhibits ADP-induced platelet aggregation in rats at 12.5 mg/kg; prevents monocrotaline-induced increases in MAP, expression of IL-1β and TNF in plasma and lung, and endothelial cell apoptosis in pulmonary arterioles in a rat model of pulmonary hypertension; reduces proteinuria and renal macrophage infiltration in a rat model of streptozotocin-induced diabetes; decreases gene expression of inflammatory cytokines, dyslipidemia, and liver steatosis in a hamster model of high-fat diet-induced NASH