Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ophthalmic acid | 495-27-2 | 99.9% | 50 MG
SDP

Catalog No. 5000341524
Encompass_Preferred

Ophthalmic acid is an analogue of GSH and serves as a marker of oxidative stress and hepatic GSH consumption. It is also identified as an inhibitor of the Glyoxalase I reaction.

  • GSH analogue
  • Marker of oxidative stress
  • Marker of hepatic GSH consumption
  • Inhibitor of Glyoxalase I reaction
  • Molecular weight: 289.29
  • Formula: C11H19N3O6
  • Appearance: Solid
  • Color: White to off-white
  • Initial source: Endogenous metabolite, animal kingdom
  • Documentation available: Data sheet, COA, SDS, handling instructions, HNMR, RP-HPLC, LCMS

Catalog No. 50-003-41524 Supplier Medchemexpress LLC Supplier No. HY12675250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Ophthalmic acid is an analogue of GSH and serves as a marker of oxidative stress and hepatic GSH consumption. It is also identified as an inhibitor of the Glyoxalase I reaction.

  • GSH analogue
  • Marker of oxidative stress
  • Marker of hepatic GSH consumption
  • Inhibitor of Glyoxalase I reaction
  • Molecular weight: 289.29
  • Formula: C11H19N3O6
  • Appearance: Solid
  • Color: White to off-white
  • Initial source: Endogenous metabolite, animal kingdom
  • Documentation available: Data sheet, COA, SDS, handling instructions, HNMR, RP-HPLC, LCMS

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.