Learn More
Cayman Chemical Ophthalmic Acid (trifluoroacetate salt), Purity: ≥98%, CAS: 495-27-2, 5 mg
SDP
Catalog No. 502913302
Encompass_Preferred
A glutathione analog and biomarker of hepatic GSH depletion; formed via the GSH synthesis pathway, substituting 2-aminobutyrate for cysteine; plasma and hepatic levels are increased in fasted mice and in a mouse model of acetaminophen-induced hepatotoxicity
Catalog No. 50-291-3302 Supplier Cayman Chemical Supplier No. 353435 MG
A glutathione analog and biomarker of hepatic GSH depletion; formed via the GSH synthesis pathway, substituting 2-aminobutyrate for cysteine; plasma and hepatic levels are increased in fasted mice and in a mouse model of acetaminophen-induced hepatotoxicity