Learn More
Med Vet International Ophthalmic Implant 20mm, J1244F, Each
Catalog No. 5000581249
Encompass_Preferred
Clients value their pets' appearance, shouldn't their veterinarian? The surgical enucleation of a pet's eye is a disheartening procedure. The pet owner is often more traumatized than the pet itself. Part of the surpeon's responsibility is creating the best possible post-operative appearance. The most common type of procedure is the transpalpebral approach. This technique requires the placement of an orbital implant to look cosmetically pleasing. With no implant, there will be a marked indentation as the potentially vacant space contracts. The Ophthalmic Implants are: - Simple to use by any surgeon - Quality silicone from Dow Corning - Lightweight and easily sterilized by autoclaving - Inexpensive implant cost - Black glossy color - Multitude of sizes - Manufactured in the U.S.A. at an FDA inspected facility Sizing guideline: measure the distance across the cornea then subtract 2mm. 20mm
Catalog No. 50-005-81249 Supplier Med Vet International Supplier No. JOR1244F
Clients value their pets' appearance, shouldn't their veterinarian? The surgical enucleation of a pet's eye is a disheartening procedure. The pet owner is often more traumatized than the pet itself. Part of the surpeon's responsibility is creating the best possible post-operative appearance. The most common type of procedure is the transpalpebral approach. This technique requires the placement of an orbital implant to look cosmetically pleasing. With no implant, there will be a marked indentation as the potentially vacant space contracts. The Ophthalmic Implants are: - Simple to use by any surgeon - Quality silicone from Dow Corning - Lightweight and easily sterilized by autoclaving - Inexpensive implant cost - Black glossy color - Multitude of sizes - Manufactured in the U.S.A. at an FDA inspected facility Sizing guideline: measure the distance across the cornea then subtract 2mm. 20mm