Learn More
McKesson General Medical Alcon CONSTELLATION Ophthalmology Tray, Viscous Fluid Control, EnGauge RFID Technology, 6 Per Box
Catalog No. NC2944096
Encompass
The Alcon CONSTELLATION Ophthalmology Tray is a sterile procedure pack designed for use in ophthalmic microsurgery. It offers precise viscous fluid control, enhancing surgical efficiency. Compatible with the Alcon CONSTELLATION Vision System, it features EnGauge RFID technology for automatic device recognition, streamlining workflows in operating rooms. This comprehensive pack includes various syringes and cannulas, providing everything needed for vitreoretinal and anterior segment procedures.
This item is not returnable. View return policy
Catalog No. NC2944096 Supplier McKesson General Medical Supplier No. 1050613
The Alcon CONSTELLATION Ophthalmology Tray is a sterile procedure pack designed for use in ophthalmic microsurgery. It offers precise viscous fluid control, enhancing surgical efficiency. Compatible with the Alcon CONSTELLATION Vision System, it features EnGauge RFID technology for automatic device recognition, streamlining workflows in operating rooms. This comprehensive pack includes various syringes and cannulas, providing everything needed for vitreoretinal and anterior segment procedures.