Learn More
Cliawaived Inc Single Dip Stick Opiate (OPI 300) Tests
SDP
Catalog No. NC1859182
Encompass_Preferred
(50 Tests)CLIA waived single urine dip stick test for Opiate. Read results from 5-60 minutes. New hermetically sealed plastic housing prevent dip card from flooding. Easy to read. Negative results as soon as 1 minute. Offers improved accuracy and sensitivity testing.
This item is not returnable. View return policy
Catalog No. NC1859182 Supplier Cliawaived Inc Supplier No. CLIAIDTCII13
(50 Tests)CLIA waived single urine dip stick test for Opiate. Read results from 5-60 minutes. New hermetically sealed plastic housing prevent dip card from flooding. Easy to read. Negative results as soon as 1 minute. Offers improved accuracy and sensitivity testing.