Learn More
Enzo Life Sciences SCREEN-WELL Opioid ligand library (100µl/well)
SDP
Catalog No. 502010194
Encompass_Preferred
The Opioid and Sigma Ligands Plate contains 74 ligands including endogenous neurotransmitters, agonists, antagonists, and marketed drugs. The library is ideal for screening or identifying recombinant orphan G protein coupled receptors, target validation, secondary screening, assay development, and other pharmacological applications. The Opioid and Sigma Ligands plate is 1 plate of the larger Neurotransmitter Library (BML-2810) which contains over 650 ligands. Long Term Storage: -80°C. Applications: HTS .
Catalog No. 50-201-0194 Supplier Enzo Life Sciences Supplier No. BML28140100
The Opioid and Sigma Ligands Plate contains 74 ligands including endogenous neurotransmitters, agonists, antagonists, and marketed drugs. The library is ideal for screening or identifying recombinant orphan G protein coupled receptors, target validation, secondary screening, assay development, and other pharmacological applications. The Opioid and Sigma Ligands plate is 1 plate of the larger Neurotransmitter Library (BML-2810) which contains over 650 ligands. Long Term Storage: -80°C. Applications: HTS .