Learn More
AdipoGen Opipramol
SDP
Supplier:  AdipoGen CDXO0042M010
Encompass_Preferred
Chemical. CAS 315-72-0. Formula C23H29N3O. MW 363.5. Synthetic. Tricyclic antidepressant and anxiolytic compound. Acts as a high affinity sigma receptor agonist, primarily at the sigma1 subtype, but also at the sigma2 subtype with somewhat lower affinity. Shows a low to moderate antagonistic affinity for the D2, 5-HT2, H1, H2, and muscarinic acetylcholine receptors. H1 and H2 receptor antagonism account for its antihistamine effects, and muscarinic acetylcholine receptor antagonism is responsible for its anticholinergic properties.
Catalog No. 50-256-0333