Learn More
Medchemexpress LLC Opiranserin hydrochloride | 1440796-75-7 | 99.6% | 430.97 | 1 ML
SDP
Catalog No. 5000344316
Encompass_Preferred
Opiranserin (VVZ-149) hydrochloride is a non-opioid and non-NSAID analgesic candidate. It acts as a dual antagonist of glycine transporter type 2 (GlyT2) and serotonin receptor 2A (5HT2A). It also shows antagonistic activity on rP2X3. Opiranserin hydrochloride is being developed as an injectable agent for the treatment of postoperative pain.
  • Is a non-opioid and non-NSAID analgesic candidate.
  • Acts as a dual antagonist of glycine transporter type 2 (GlyT2) and serotonin receptor 2A (5HT2A).
  • Exhibits antagonistic activity on rP2X3.
  • Is being developed as an injectable agent for the treatment of postoperative pain.
Catalog No. 50-003-44316 Supplier Medchemexpress LLC Supplier No. HY109067A10MG
Opiranserin (VVZ-149) hydrochloride is a non-opioid and non-NSAID analgesic candidate. It acts as a dual antagonist of glycine transporter type 2 (GlyT2) and serotonin receptor 2A (5HT2A). It also shows antagonistic activity on rP2X3. Opiranserin hydrochloride is being developed as an injectable agent for the treatment of postoperative pain.
  • Is a non-opioid and non-NSAID analgesic candidate.
  • Acts as a dual antagonist of glycine transporter type 2 (GlyT2) and serotonin receptor 2A (5HT2A).
  • Exhibits antagonistic activity on rP2X3.
  • Is being developed as an injectable agent for the treatment of postoperative pain.