Learn More
Medchemexpress LLC Opiranserin hydrochloride | 1440796-75-7 | 99.6% | 430.97 | 10 MG
SDP
Catalog No. 5000345474
Encompass_Preferred
Opiranserin hydrochloride is a non-opioid and non-NSAID analgesic candidate. It functions as a dual antagonist of glycine transporter type 2 (GlyT2) and serotonin receptor 2A (5HT2A). It also exhibits antagonistic activity on rP2X3. Opiranserin hydrochloride is being developed as an injectable agent for the treatment of postoperative pain.
  • Functions as a dual antagonist of GlyT2 and 5HT2A.
  • Exhibits antagonistic activity on rP2X3.
  • Effectively reduces mechanical allodynia and pain-related behaviors.
  • Involved in clinical trials for postoperative pain, healthy, and radiculopathy/sciatica.
Catalog No. 50-003-45474 Supplier Medchemexpress LLC Supplier No. HY109067A100MG
Opiranserin hydrochloride is a non-opioid and non-NSAID analgesic candidate. It functions as a dual antagonist of glycine transporter type 2 (GlyT2) and serotonin receptor 2A (5HT2A). It also exhibits antagonistic activity on rP2X3. Opiranserin hydrochloride is being developed as an injectable agent for the treatment of postoperative pain.
  • Functions as a dual antagonist of GlyT2 and 5HT2A.
  • Exhibits antagonistic activity on rP2X3.
  • Effectively reduces mechanical allodynia and pain-related behaviors.
  • Involved in clinical trials for postoperative pain, healthy, and radiculopathy/sciatica.