Learn More
Medchemexpress LLC Opiranserin hydrochloride | 1440796-75-7 | MFCD34676620 | 99.6% | 430.97 g/mol | C21H35ClN2O5 | 5 MG
SDP
Catalog No. 5000264600
Encompass_Preferred
Opiranserin hydrochloride is a research-stage, non-opioid analgesic candidate that acts as a dual antagonist of glycine transporter 2 (GlyT2) and the serotonin 2A receptor (5-HT2A). Supplied as the hydrochloride salt, it is used in preclinical pharmacology and mechanism-of-action studies.
  • Dual antagonist activity at GlyT2 and 5-HT2A demonstrated in vitro.
  • Non-opioid, non-NSAID analgesic candidate suitable for postoperative pain research.
  • Available in solid and DMSO solution formats for flexible dosing and formulation.
  • High reported purity (99.6%) appropriate for preclinical experiments.
  • Supplied in small pack sizes compatible with early-stage studies.
Catalog No. 50-002-64600 Supplier Medchemexpress LLC Supplier No. HY109067A5MG
Opiranserin hydrochloride is a research-stage, non-opioid analgesic candidate that acts as a dual antagonist of glycine transporter 2 (GlyT2) and the serotonin 2A receptor (5-HT2A). Supplied as the hydrochloride salt, it is used in preclinical pharmacology and mechanism-of-action studies.
  • Dual antagonist activity at GlyT2 and 5-HT2A demonstrated in vitro.
  • Non-opioid, non-NSAID analgesic candidate suitable for postoperative pain research.
  • Available in solid and DMSO solution formats for flexible dosing and formulation.
  • High reported purity (99.6%) appropriate for preclinical experiments.
  • Supplied in small pack sizes compatible with early-stage studies.