Learn More
Medchemexpress LLC Opiranserin hydrochloride (VVZ-149 hydrochloride) | 1440796-75-7 | 99.6% | 430.97 | 50 MG
SDP
Catalog No. 5000344377
Encompass_Preferred
Opiranserin hydrochloride (VVZ-149 hydrochloride) is a non-opioid and non-NSAID analgesic candidate. It functions as a dual antagonist of glycine transporter type 2 (GlyT2) and serotonin receptor 2A (5HT2A), with IC50 values of 0.86 and 1.3 μM, respectively. Additionally, it demonstrates antagonistic activity on rP2X3 (IC50=0.87 μM). It is being developed as an injectable agent for the treatment of postoperative pain and is intended for research use only.
  • Functions as dual antagonist of GlyT2 and 5HT2A
  • Demonstrates antagonistic activity on rP2X3
  • Intended for research use only
  • Developed as an injectable agent for postoperative pain
Catalog No. 50-003-44377 Supplier Medchemexpress LLC Supplier No. HY109067A10MM/1ML
Opiranserin hydrochloride (VVZ-149 hydrochloride) is a non-opioid and non-NSAID analgesic candidate. It functions as a dual antagonist of glycine transporter type 2 (GlyT2) and serotonin receptor 2A (5HT2A), with IC50 values of 0.86 and 1.3 μM, respectively. Additionally, it demonstrates antagonistic activity on rP2X3 (IC50=0.87 μM). It is being developed as an injectable agent for the treatment of postoperative pain and is intended for research use only.
  • Functions as dual antagonist of GlyT2 and 5HT2A
  • Demonstrates antagonistic activity on rP2X3
  • Intended for research use only
  • Developed as an injectable agent for postoperative pain