Learn More
Macherey-Nagel GC column OPTIMA 1, 0.53 mm ID, 2.00 µm, 50 m
Catalog No. NC0280347
Encompass_Preferred
Capillary column OPTIMA 1 - ID: 0.53 mm, film thickness: 2.00 µm - length: 50 m - pack of 1
Catalog No. NC0280347 Supplier Macherey-Nagel Supplier No. 726521.50
Capillary column OPTIMA 1 - ID: 0.53 mm, film thickness: 2.00 µm - length: 50 m - pack of 1