Learn More
Enzo Life Sciences Osteopontin monoclonal antibody (53) (100 µg)
SDP
Supplier:  Enzo Life Sciences ADI905629100
Encompass_Preferred
Clone: 53. Host: Mouse. Isotype: IgG2a. Immunogen: Recombinant human Osteopontin. UniProt: P10451. Source: Purified from ascites. Species Reactivity: Human. Applications: ELISA, IHC (FS), IHC (PS), WB. Purity: Protein G affinity purified. Formulation: Liquid. In PBS containing 0.1% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-658-386