Learn More
Aviva Systems Biology Corp OPN1MW Antibody - C-terminal region (ARP59764_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59764_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against OPN1MW. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human OPN1MW. Alternate names: CBBM, CBD, GCP, MGC176615, MGC177321, MGC198468, MGC198469, OPN1MW1, OPN1MW2, GOP, COD5ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-286-816