Learn More
Aviva Systems Biology Corp Opn1mw Antibody - C-terminal region (ARP59775_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59775_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Opn1mw. It was validated on Western Blot. . Alternate names: GcpApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-820