Learn More
Biomatik Corporation Opn1Mw Polyclonal Antibody
Supplier:  Biomatik Corporation CAC11491
Encompass_Preferred
Opn1Mw Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to OPN1MW. The Opn1Mw Polyclonal Antibody has been validated for the following applications: ELISA, WB, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Medium-Wave-Sensitive Opsin 1 Protein (1-52Aa). Applications: ELISA, WB, IF. Target Synonyms: OPN1MW. Accession Number: P04001. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6701