Learn More
Aviva Systems Biology Corp OPN1SW Antibody - C-terminal region (ARP59911_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59911_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against OPN1SW. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human OPN1SW. Alternate names: BCP, BOP, CBTApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-874