Learn More
Aviva Systems Biology Corp OPN3 Antibody - C-terminal region (ARP59791_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59791_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against OPN3. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human OPN3. Alternate names: ECPNApplicationWB. Species reactivity: Guinea Pig, Horse, Human
Catalog No. 50-286-826