Learn More
Aviva Systems Biology Corp Opn3 Antibody - C-terminal region (ARP59792_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59792_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Opn3. It was validated on Western Blot. . Alternate names: ERO, Ecpn, MGC124138, panopsinApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-827