Learn More
Grainger ITL Insulated Open Ended Spanner, 10mm, Alloy Steel, 1 ea
Catalog No. 502783053
Encompass_Preferred
This 10 mm insulated open-ended spanner is engineered for electrical and live line protection. Manufactured from 31CrV3 alloy steel with a unique injection-molded nylon thermoplastic insulation, this tool provides a secure, bonded interface between the metal and insulation for enhanced durability.
  • Accredited to IEC 60900:2018 and ASTM F1505 standards
  • Category C impact tested for operations in temperatures from -40°C to +70°C
  • Flame retardant and resistant to extreme impact and abrasion
  • Tested at 10,000 volts for 3 minutes submerged in water
  • Harder and more durable than PVC coated alternatives
  • Lifetime warranty on tool and insulation
Catalog No. 50-278-3053 Supplier Grainger Supplier No. 19MU97
This 10 mm insulated open-ended spanner is engineered for electrical and live line protection. Manufactured from 31CrV3 alloy steel with a unique injection-molded nylon thermoplastic insulation, this tool provides a secure, bonded interface between the metal and insulation for enhanced durability.
  • Accredited to IEC 60900:2018 and ASTM F1505 standards
  • Category C impact tested for operations in temperatures from -40°C to +70°C
  • Flame retardant and resistant to extreme impact and abrasion
  • Tested at 10,000 volts for 3 minutes submerged in water
  • Harder and more durable than PVC coated alternatives
  • Lifetime warranty on tool and insulation