Learn More
Grainger ITL Open End Wrench, Alloy Steel, 1/2 in, Insulated, 1 ea
Catalog No. 502782795
Encompass_Preferred
This insulated open-end wrench is a professional-grade tool designed for safe operation in electrical environments. Engineered for durability and high performance, the tool features a 1/2 in. head size and is constructed from 31CrV3 steel alloy, finished with chrome for corrosion resistance. The one-piece injection-molded nylon thermoplastic insulation ensures maximum safety and longevity, surpassing the performance of standard dipped tools.
  • Rated for 1,000V AC and tested to 10,000V
  • Meets IEC 60900:2018 and ASTM F1505 standards
  • Category C impact tested for temperature extremes from -40°C to +70°C
  • Integrated 15° head offset for improved access in confined areas
  • Features warning thumb stops and a fixed head design
  • Lifetime warranty on both the tool and its insulation
Catalog No. 50-278-2795 Supplier Grainger Supplier No. 19MV03
This insulated open-end wrench is a professional-grade tool designed for safe operation in electrical environments. Engineered for durability and high performance, the tool features a 1/2 in. head size and is constructed from 31CrV3 steel alloy, finished with chrome for corrosion resistance. The one-piece injection-molded nylon thermoplastic insulation ensures maximum safety and longevity, surpassing the performance of standard dipped tools.
  • Rated for 1,000V AC and tested to 10,000V
  • Meets IEC 60900:2018 and ASTM F1505 standards
  • Category C impact tested for temperature extremes from -40°C to +70°C
  • Integrated 15° head offset for improved access in confined areas
  • Features warning thumb stops and a fixed head design
  • Lifetime warranty on both the tool and its insulation