Learn More
Grainger Insulated Tools US 13 mm Open End Wrench, Nylon Thermoplastic, 1,000V, 1 ea
Catalog No. 502779426
Encompass_Preferred
The 13 mm insulated open ended wrench is an injection-molded, heavy-duty tool engineered for live line working. Crafted from high-strength steel alloy, it features an integrated nylon thermoplastic insulation that is bonded as one with the metal, ensuring durability and flame resistance. This tool is designed to provide superior performance in harsh environments and extreme temperatures.
  • Meets IEC 60900:2018 and ASTM F1505 industry standards
  • Rated for operation in temperatures ranging from -40°C to +70°C
  • Tested at 10,000 V for 3 minutes while submerged in water
  • Features a 15-degree head angle for enhanced access
  • Constructed from steel alloy 31CrV3 with impact and abrasion resistance
  • Includes a lifetime warranty on both the tool and insulation
Catalog No. 50-277-9426 Supplier Grainger Supplier No. 19MU98
The 13 mm insulated open ended wrench is an injection-molded, heavy-duty tool engineered for live line working. Crafted from high-strength steel alloy, it features an integrated nylon thermoplastic insulation that is bonded as one with the metal, ensuring durability and flame resistance. This tool is designed to provide superior performance in harsh environments and extreme temperatures.
  • Meets IEC 60900:2018 and ASTM F1505 industry standards
  • Rated for operation in temperatures ranging from -40°C to +70°C
  • Tested at 10,000 V for 3 minutes while submerged in water
  • Features a 15-degree head angle for enhanced access
  • Constructed from steel alloy 31CrV3 with impact and abrasion resistance
  • Includes a lifetime warranty on both the tool and insulation