Learn More
Grainger ITL Insulated Open Ended Spanner, Metric, 17 mm, 1 ea
Catalog No. 502783054
Encompass_Preferred
This insulated open ended spanner is designed for electrical work, providing safety and reliability. Constructed from durable chrome-vanadium steel, the tool is fully insulated and features a fixed, 15-degree head angle for improved access in confined spaces.
  • Features a 17 mm head size
  • Constructed from durable chrome-vanadium steel
  • Equipped with an orange, one-piece injection-molded handle
  • Corrosion-resistant finish
  • Certified to IEC 60900:2018 and ASTM F1505 standards
  • Backed by a lifetime guarantee on both the tool and insulation
Catalog No. 50-278-3054 Supplier Grainger Supplier No. 19MU99
This insulated open ended spanner is designed for electrical work, providing safety and reliability. Constructed from durable chrome-vanadium steel, the tool is fully insulated and features a fixed, 15-degree head angle for improved access in confined spaces.
  • Features a 17 mm head size
  • Constructed from durable chrome-vanadium steel
  • Equipped with an orange, one-piece injection-molded handle
  • Corrosion-resistant finish
  • Certified to IEC 60900:2018 and ASTM F1505 standards
  • Backed by a lifetime guarantee on both the tool and insulation