Learn More
Grainger Insulated Tools US Open End Wrench, 3/4 In., 1000V, SAE, 1 ea.
Catalog No. 502783220
Encompass_Preferred
This 3/4 in. open ended wrench is designed for safe use in live line working environments. Engineered for durability and reliability, it features a one-piece injection-molded nylon thermoplastic insulation bonded to the steel alloy tool, providing superior strength and hardness compared to standard PVC-coated alternatives. Each tool undergoes 100% testing at 10,000 volts to ensure safety and performance.
  • Meets IEC 60900:2018 and ASTM F1505 standards for live line working
  • Category C impact tested for operation in extreme temperatures from -40°C to +70°C (-40°F to +158°F)
  • Features a 15° head angle for improved access in confined spaces
  • Constructed from high-quality 31CrV3 alloy steel with a corrosion-resistant finish
  • Flame retardant, impact, and abrasion resistant design
  • Lifetime warranty on both the tool and insulation
Catalog No. 50-278-3220 Supplier Grainger Supplier No. 19MV05
This 3/4 in. open ended wrench is designed for safe use in live line working environments. Engineered for durability and reliability, it features a one-piece injection-molded nylon thermoplastic insulation bonded to the steel alloy tool, providing superior strength and hardness compared to standard PVC-coated alternatives. Each tool undergoes 100% testing at 10,000 volts to ensure safety and performance.
  • Meets IEC 60900:2018 and ASTM F1505 standards for live line working
  • Category C impact tested for operation in extreme temperatures from -40°C to +70°C (-40°F to +158°F)
  • Features a 15° head angle for improved access in confined spaces
  • Constructed from high-quality 31CrV3 alloy steel with a corrosion-resistant finish
  • Flame retardant, impact, and abrasion resistant design
  • Lifetime warranty on both the tool and insulation