Learn More
Biomatik Corporation Oprf Polyclonal Antibody
Supplier:  Biomatik Corporation CAC11605
Encompass_Preferred
Oprf Polyclonal Antibody is a Pseudomonas aeruginosa Polyclonal Antibody to oprF. The Oprf Polyclonal Antibody has been validated for the following applications: ELISA, WB. Host: Rabbit. Species: Pseudomonas aeruginosa. Immunogen: Recombinant Pseudomonas Aeruginosa Outer Membrane Porin F Protein (25-350Aa). Applications: ELISA, WB. Target Synonyms: oprF. Accession Number: P13794. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9239