Learn More
Aviva Systems Biology Corp OPRK1 Antibody - N-terminal region (AVARP13050_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP13050_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against OPRK1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human OPRK1. Alternate names: KOR, OPRKApplicationWB. Species reactivity: Guinea Pig, Human, Mouse, Pig
Catalog No. 50-014-62624