Learn More
Aviva Systems Biology Corp OPRL1 Antibody - middle region (ARP59570_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59570_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against OPRL1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human OPRL1. Alternate names: KOR-3, MGC34578, NOCIR, OOR, ORL1ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-286-744