Learn More
Cayman Chemical OPropargylPuromycIn Stock25uL
SDP
Catalog No. 502895657
Encompass_Preferred
A GLP-1R positive allosteric modulator enhances GLP-1 binding to GLP-1R (EC50 11.01 UM) and cAMP accumulation induced by GLP-1 (7-36) in CHO cells expressing GLP-1R
Catalog No. 50-289-5657 Supplier Cayman Chemical Supplier No. 60110125 UL
A GLP-1R positive allosteric modulator enhances GLP-1 binding to GLP-1R (EC50 11.01 UM) and cAMP accumulation induced by GLP-1 (7-36) in CHO cells expressing GLP-1R