Learn More
Medchemexpress LLC HY-12113 10mg , Oprozomib CAS:935888-69-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1211310MG
Encompass_Preferred
HY-12113 10mg Oprozomib CAS: 935888-69-0 Oprozomib (PR-047) is an orally bioavailable and selective peptide epoxyketone proteasome inhibitor with IC50s of 36 and 82 nM for proteasome (β5) and immunoproteasome (LMP7), respectively. Oprozomib (ONX 0912) induces apoptosis in MM cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0606