Learn More
Apexbio Technology LLC Oprozomib (ONX-0912) 935888-69-0 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062787
Encompass_Preferred
Oprozomib (ONX-0912 CAS 935888-69-0) is an orally bioavailable small-molecule inhibitor of the proteasome structurally related to carfilzomib It targets predominantly the chymotrypsin-like proteolytic activity of the proteasome inducing apoptosis and growth inhibition in multiple myeloma (MM) cell lines including those resistant to bortezomib In preclinical models oprozomib activates pro-apoptotic pathways involving caspases-3 -8 -9 as well as PARP cleavage suppresses angiogenesis and tumor cell migration thereby reducing tumor progression and prolonging survival Its biological activity positions oprozomib as a promising compound for MM research
Catalog No. 50-000-62787 Supplier Apexbio Technology LLC Supplier No. A19345.1
Oprozomib (ONX-0912 CAS 935888-69-0) is an orally bioavailable small-molecule inhibitor of the proteasome structurally related to carfilzomib It targets predominantly the chymotrypsin-like proteolytic activity of the proteasome inducing apoptosis and growth inhibition in multiple myeloma (MM) cell lines including those resistant to bortezomib In preclinical models oprozomib activates pro-apoptotic pathways involving caspases-3 -8 -9 as well as PARP cleavage suppresses angiogenesis and tumor cell migration thereby reducing tumor progression and prolonging survival Its biological activity positions oprozomib as a promising compound for MM research