Learn More
Agilent Technologies (OPS REAGENT) O-PHTHALAL-6X1ML
Catalog No. NC2164156
Encompass
(OPS Reagent) o-phthalaldehyde & 3-mercaptopropionic acid in borate buffer
This item is not returnable. View return policy
Catalog No. NC2164156 Supplier Agilent Technologies Supplier No. 50613335
(OPS Reagent) o-phthalaldehyde & 3-mercaptopropionic acid in borate buffer