Learn More
EMD MILLIPORE 3P BIOPHARM OPTCP GMMA CMPTBL XL CPSL FLTR
Supplier:  EMD MILLIPORE 3P BIOPHARM KHGEG003HB3
Encompass
Opticap Gamma Compatible XL300 Millipore Express SHC 0.5 + 0.2um 9/16in. HB/HB with Bell
Catalog No. NC0598456