Learn More
Optimize Technologies OPTI-GUARD™ 1mm Guard Column, C4, 5pk
SDP
Catalog No. 502006099
Encompass_Preferred
OPTI-GUARD™ sets the standard for extremely low impact, easy to use pre-column protection. Designed for use with analytical (4.6, 3.0 mm i.d.) and narrow-bore (2.1, 1.0 mm i.d.) columns, the patented floating stem design automatically adjusts to any manufacturer's tube stop depth for a zero-dead-volume connection every time. The best part? It's no larger than a standard fitting. Connecting the OPTI-GUARD™ requires no extra tubing, maintaining the minimal impact philosophy that has made the OPTI-GUARD™ line so popular. As always, custom packing is available. Key Features include: 5 disposable hand-tight holders and cartridges, a ZDV Connection, automatic adjusting to any manufacturer's column, no extra tubing is required for connection, the product does not effect chromatography, and it is no larger than a standard fitting.
Catalog No. 50-200-6099 Supplier Optimize Technologies Supplier No. 100203211
OPTI-GUARD™ sets the standard for extremely low impact, easy to use pre-column protection. Designed for use with analytical (4.6, 3.0 mm i.d.) and narrow-bore (2.1, 1.0 mm i.d.) columns, the patented floating stem design automatically adjusts to any manufacturer's tube stop depth for a zero-dead-volume connection every time. The best part? It's no larger than a standard fitting. Connecting the OPTI-GUARD™ requires no extra tubing, maintaining the minimal impact philosophy that has made the OPTI-GUARD™ line so popular. As always, custom packing is available. Key Features include: 5 disposable hand-tight holders and cartridges, a ZDV Connection, automatic adjusting to any manufacturer's column, no extra tubing is required for connection, the product does not effect chromatography, and it is no larger than a standard fitting.