Learn More
Abcam Opti-Klear™ Live Cell Imaging Buffer 1 x , 60ML
Catalog No. NC1835081
Encompass_Preferred
Hepes based imaging buffer formulated to lower background fluorescence and maintain proper pH and osmolarity for hours at 37°C. ab275938 provides an energy source and preserves fluorescent signals.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1835081 Supplier Abcam Supplier No. AB27593860ML
Hepes based imaging buffer formulated to lower background fluorescence and maintain proper pH and osmolarity for hours at 37°C. ab275938 provides an energy source and preserves fluorescent signals.
The product is subject to the following: Abcam Restricted Use Statement