Learn More
Optimize Technologies OPTI-LOK™ 6K 1/4-28 SSI Style Male Nut, PEEK
SDP
Catalog No. 50113347
Encompass_Preferred
OPTI-LOK Fittings are ideal for high-pressure connections with 1/16” OD tubing. All Optimize fittings are precision machined for optimal quality and thread consistency and are designed for trouble-free operation at pressures of up to 6,000 psi.
Catalog No. 50-113-347 Supplier Optimize Technologies Supplier No. 102000330
OPTI-LOK Fittings are ideal for high-pressure connections with 1/16” OD tubing. All Optimize fittings are precision machined for optimal quality and thread consistency and are designed for trouble-free operation at pressures of up to 6,000 psi.