Learn More
Optimize Technologies OPTI-LOK™ 6K 1/4-28 SSI Style Male Nut & PEEK Ferrules, 10pk (10-20-00330 & 10-20-00269)
SDP
Catalog No. 50113348
Encompass_Preferred
OPTI-LOK Fittings are ideal for high-pressure connections with 1/16” OD tubing. All Optimize fittings are precision machined for optimal quality and thread consistency and are designed for trouble-free operation at pressures of up to 6,000 psi.
Catalog No. 50-113-348 Supplier Optimize Technologies Supplier No. 102000331
OPTI-LOK Fittings are ideal for high-pressure connections with 1/16” OD tubing. All Optimize fittings are precision machined for optimal quality and thread consistency and are designed for trouble-free operation at pressures of up to 6,000 psi.