Learn More
Optimize Technologies OPTI-LOK™ 6K Hex Head Short Nuts & Ferrules, 10pk (10-20-00271 & 10-20-00269)
SDP
Catalog No. 50113336
Encompass_Preferred
OPTI-LOK™ Fittings are ideal for high-pressure connections with 1/16” OD tubing. All Optimize fittings are precision machined for optimal quality and thread consistency and are designed for trouble-free operation at pressures of up to 6,000 psi.
Catalog No. 50-113-336 Supplier Optimize Technologies Supplier No. 102000272
OPTI-LOK™ Fittings are ideal for high-pressure connections with 1/16” OD tubing. All Optimize fittings are precision machined for optimal quality and thread consistency and are designed for trouble-free operation at pressures of up to 6,000 psi.