Learn More
Optimize Technologies OPTI-LOK™ 6K PEEK Union, .010" Thru
SDP
Catalog No. 50113325
Encompass_Preferred
OPTI-LOK™ Fittings are ideal for high-pressure connections with 1/16” OD tubing. All Optimize fittings are precision machined for optimal quality and thread consistency and are designed for trouble-free operation at pressures of up to 6,000 psi. The OPTI-LOK™ high pressure unions offer 10-32 threads for all high pressure applications with 1/16" OD tubing and provide zero dead volume connections with every use.
Catalog No. 50-113-325 Supplier Optimize Technologies Supplier No. 102000261
OPTI-LOK™ Fittings are ideal for high-pressure connections with 1/16” OD tubing. All Optimize fittings are precision machined for optimal quality and thread consistency and are designed for trouble-free operation at pressures of up to 6,000 psi. The OPTI-LOK™ high pressure unions offer 10-32 threads for all high pressure applications with 1/16" OD tubing and provide zero dead volume connections with every use.