Learn More
Optimize Technologies OPTI-LOK™ Adapter, Luer Female to 10-32 Parker Female, PEEK
SDP
Catalog No. 50113411
Encompass_Preferred
OPTI-LOK Fittings are ideal for high-pressure connections with 1/16” OD tubing. All Optimize fittings are precision machined for optimal quality and thread consistency and are designed for trouble-free operation at pressures of up to 6,000 psi.
Catalog No. 50-113-411 Supplier Optimize Technologies Supplier No. 102300358
OPTI-LOK Fittings are ideal for high-pressure connections with 1/16” OD tubing. All Optimize fittings are precision machined for optimal quality and thread consistency and are designed for trouble-free operation at pressures of up to 6,000 psi.