Learn More
Optimize Technologies OPTI-LOK™ Coupler, PEEK
SDP
Catalog No. 50113330
Encompass_Preferred
OPTI-LOK™ Fittings are ideal for high-pressure connections with 1/16” OD tubing. All Optimize fittings are precision machined for optimal quality and thread consistency and are designed for trouble-free operation at pressures of up to 6,000 psi. OPTI-LOK™ 10-32 Couplers include (2) OPTI-LOK I fittings with integral ferrules and 6cm of tubing.
Catalog No. 50-113-330 Supplier Optimize Technologies Supplier No. 102000266
OPTI-LOK™ Fittings are ideal for high-pressure connections with 1/16” OD tubing. All Optimize fittings are precision machined for optimal quality and thread consistency and are designed for trouble-free operation at pressures of up to 6,000 psi. OPTI-LOK™ 10-32 Couplers include (2) OPTI-LOK I fittings with integral ferrules and 6cm of tubing.