Learn More
Optimize Technologies OPTI-LOK™ I, Hand Tight Fitting with Integral Ferrule, PCTFE, 10pk
SDP
Catalog No. 50113324
Encompass_Preferred
OPTI-LOK™ Fittings are ideal for high-pressure connections with 1/16” OD tubing. All Optimize fittings are precision machined for optimal quality and thread consistency and are designed for trouble-free operation at pressures of up to 6,000 psi.
Catalog No. 50-113-324 Supplier Optimize Technologies Supplier No. 102000259
OPTI-LOK™ Fittings are ideal for high-pressure connections with 1/16” OD tubing. All Optimize fittings are precision machined for optimal quality and thread consistency and are designed for trouble-free operation at pressures of up to 6,000 psi.