Learn More
Optimize Technologies OPTI-LOK™ Low Pressure 1/4-28 Plug, PTFE
SDP
Catalog No. 50113382
Encompass_Preferred
OPTI-LOK™ Low Pressure Fittings are an excellent choice for making low-pressure connections into 1/4-28 ports. Connections to flat-bottomed ports can be made using OPTI-LOK™ flat-bottomed ferrules for 1/16” or 1/8” tubing without the need for pre-flanged tubing ends. OPTI-LOK™ Low Pressure Fittings are precision machined to deliver leak-free, zero-dead-volume connections every time. Optimize also offers 1/4-28 unions in various through-hole sizes for joining flat-bottom fittings.
Catalog No. 50-113-382 Supplier Optimize Technologies Supplier No. 102100326
OPTI-LOK™ Low Pressure Fittings are an excellent choice for making low-pressure connections into 1/4-28 ports. Connections to flat-bottomed ports can be made using OPTI-LOK™ flat-bottomed ferrules for 1/16” or 1/8” tubing without the need for pre-flanged tubing ends. OPTI-LOK™ Low Pressure Fittings are precision machined to deliver leak-free, zero-dead-volume connections every time. Optimize also offers 1/4-28 unions in various through-hole sizes for joining flat-bottom fittings.