Learn More
Optimize Technologies OPTI-LOK™ Low Pressure Union, Natural PEEK
SDP
Catalog No. 50113381
Encompass_Preferred
OPTI-LOK™ Low Pressure Fittings are an excellent choice for making low-pressure connections into 1/4-28 ports. Connections to flat-bottomed ports can be made using OPTI-LOK™ flat-bottomed ferrules for 1/16” or 1/8” tubing without the need for pre-flanged tubing ends. OPTI-LOK™ Low Pressure Fittings are precision machined to deliver leak-free, zero-dead-volume connections every time. The OPTI-LOK™ low pressure unions feature 1/4-28 threads for all high pressure applications.
Catalog No. 50-113-381 Supplier Optimize Technologies Supplier No. 102100323
OPTI-LOK™ Low Pressure Fittings are an excellent choice for making low-pressure connections into 1/4-28 ports. Connections to flat-bottomed ports can be made using OPTI-LOK™ flat-bottomed ferrules for 1/16” or 1/8” tubing without the need for pre-flanged tubing ends. OPTI-LOK™ Low Pressure Fittings are precision machined to deliver leak-free, zero-dead-volume connections every time. The OPTI-LOK™ low pressure unions feature 1/4-28 threads for all high pressure applications.