Learn More
Optimize Technologies OPTI-LOK™ Plug, Natural PEEK, 10pk
SDP
Catalog No. 50113322
Encompass
We realize that your time is valuable and should not be wasted worrying about fittings. For this reason, all Optimize fittings are precision machined in the USA for optimal quality and thread consistency. The machining process is the best way to ensure tight manufacturing tolerances and minimal product variance. Optimize OPTI-LOK™ fittings are available in both low pressure and high pressure options that are sourced from various materials such as Natural PEEK, PTFE and White Acetal. This product specifically is optimal for closing off a column or other ports that are not in use.
Catalog No. 50-113-322 Supplier Optimize Technologies Supplier No. 102000256
We realize that your time is valuable and should not be wasted worrying about fittings. For this reason, all Optimize fittings are precision machined in the USA for optimal quality and thread consistency. The machining process is the best way to ensure tight manufacturing tolerances and minimal product variance. Optimize OPTI-LOK™ fittings are available in both low pressure and high pressure options that are sourced from various materials such as Natural PEEK, PTFE and White Acetal. This product specifically is optimal for closing off a column or other ports that are not in use.