Learn More
Optimize Technologies OPTI-LYNX™ Bayonet Connecter w/ 10-32 Female Port
SDP
Catalog No. 50113436
Encompass_Preferred
All OPTI-LYNX™ hardware can be purchased in kits or separately as needed for your application. OPTI-LYNX™ hardware is precision machined to maintain the highest quality and strictest tolerances. Use an OPTI-LYNX™ in-line kit & an extra bayonet connector to switch quickly and easily between multiple detectors or flow paths. Simply terminate the column outlet line with a holder cap/tube and use extra bayonet connectors for each detector inlet line. Use with other OPTI-LYNX™ hardware and easily modify your OPTI-LYNX™ holder from a "Direct Connect" to an "In-Line" version.
Catalog No. 50-113-436 Supplier Optimize Technologies Supplier No. 1102212AA
All OPTI-LYNX™ hardware can be purchased in kits or separately as needed for your application. OPTI-LYNX™ hardware is precision machined to maintain the highest quality and strictest tolerances. Use an OPTI-LYNX™ in-line kit & an extra bayonet connector to switch quickly and easily between multiple detectors or flow paths. Simply terminate the column outlet line with a holder cap/tube and use extra bayonet connectors for each detector inlet line. Use with other OPTI-LYNX™ hardware and easily modify your OPTI-LYNX™ holder from a "Direct Connect" to an "In-Line" version.